missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562827
This item is not returnable.
View return policy
Description
Immunogen sequence: NIRPGETGMD VTSRCTLGDP NKLPEGVPQP ARMPYISDKH PRQTLEVINL LRKHRELCDV VLVVGAKKIY AHRVILSA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 44-56 amino acid repeat motif. The kelch motif appears in many different polypeptide contexts and contains multiple potential protein-protein contact sites. Members of this family are present both throughout the cell and extracellularly, with diverse activities.
Specifications
| KLHL20 | |
| Polyclonal | |
| Unconjugated | |
| KLHL20 | |
| AI504637; D930050H05Rik; kelch like family member 20; Kelch motif containing protein; kelch-like 18; kelch-like 20; Kelch-like ECT2 interacting protein; kelch-like ECT2-interacting protein; kelch-like family member 20; kelch-like protein 20; Kelch-like protein X; KHLHX; Kiaa4210; Kleip; klh20; KLHL20; KLHLX; mKIAA4210; RGD1309490; RP3-383J4.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2M5 | |
| KLHL20 | |
| Recombinant protein corresponding to Human KLHL20. Recombinant protein control fragment (Product #RP-103164). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction