missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL24 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05979APC
This item is not returnable.
View return policy
Description
KLHL24 Polyclonal antibody specifically detects KLHL24 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| KLHL24 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| DRE1, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24, KRIP6, Protein DRE1 | |
| Produced in rabbits immunized with E. coli-derived Human KLHL24 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 54800 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction