missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL25 Antibody, DyLight 680, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-77147FR
This item is not returnable.
View return policy
Description
KLHL25 Polyclonal antibody specifically detects KLHL25 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| KLHL25 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 680 | |
| ectodermal-neural cortex 2, ENC-2BTB/POZ KELCH domain protein, ENC2ectoderm-neural cortex protein 2, FLJ12587, FLJ45015, kelch-like 25 (Drosophila), Kelch-like protein 25 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human ENC-2. The immunogen is located within amino acids 230 - 280 of ENC-2. Amino Acid Squence: SSEALMADERTKL | |
| 0.1 mL | |
| Neuroscience | |
| 64410 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction