missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL28 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | KLHL28 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458091
|
Novus Biologicals
NBP1-90748-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221328
|
Novus Biologicals
NBP1-90748 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLHL28 Polyclonal specifically detects KLHL28 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| KLHL28 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 54813 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QNFEAVCQTEEFFELTHADLDEIVSNDCLNVATEETVFYALESWIKYDVQERQKYLAQLLNSVRLPLLSVKFLTRLYEANHLIRDDRTCKHLLNEALKYHFMPEHRLSHQTVLMTRPRCAPKVLCAVGGKSGLFACL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BTB (POZ) domain containing 5, BTB/POZ domain-containing protein 5, FLJ20081, kelch-like 28 (Drosophila) | |
| KLHL28 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title