Learn More
KLHL28 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25224-1-AP-20UL
This item is not returnable.
View return policy
Description
BTBD5 is a transcription factor containing BTB (POZ) domain.Specifications
| KLHL28 | |
| Polyclonal | |
| Unconjugated | |
| KLHL28 | |
| BTBD5, kelch like 28 (Drosophila), Kelch like protein 28, KLHL28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54813, 66689 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9CR40, Q9NXS3 | |
| KLHL28 | |
| KLHL28 Fusion Protein Ag18665 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |