missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563834
This item is not returnable.
View return policy
Description
Immunogen sequence: DRGVVYISGG KAGRGEGGAS SLRDLYVLGP EEQVWSKKAP MGTARFGHHM AVLRGAVFAF LGRYEPFSEI ERYDPGADQW TR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
KLHL34 (Kelch Like Family Member 34) is a Protein Coding gene. An important paralog of this gene is KLHL13.
Specifications
| KLHL34 | |
| Polyclonal | |
| Unconjugated | |
| KLHL34 | |
| FLJ34960 antibody; kelch like family member 34; kelch-like 34; kelch-like 34 (Drosophila) antibody; kelch-like family member 34; kelch-like protein 34; kelch-like protein 34 antibody; KLHL34; KLHL34 antibody; MGC125650 antibody; RGD1560439; RP11-450P7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 257240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N239 | |
| KLHL34 | |
| Recombinant protein corresponding to Human KLHL34. Recombinant protein control fragment (Product #RP-101564). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction