missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554498
This item is not returnable.
View return policy
Description
Immunogen sequence: GDKSLPCHRL ILSACSPYFR EYFLSEIDEA KKKEVVLDNV DPAILDLIIK YLYSASIDLN DGNVQD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
Specifications
| KLHL41 | |
| Polyclonal | |
| Unconjugated | |
| KLHL41 | |
| Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60662 | |
| KLHL41 | |
| Recombinant protein corresponding to Human KLHL41. Recombinant protein control fragment (Product #RP-93585). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction