Learn More
KLK11 Mouse anti-Human, CoraLite 488, Clone: 1E2B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67427-100UL
Description
The KLK11 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLK11. This antibody recognizes human, pig, rat antigen. The KLK11 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| KLK11 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 11012 | |
| IgG1 |
| Immunofluorescence | |
| 1E2B4 | |
| Mouse | |
| Primary | |
| Human, Rat, Pig |