missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK3 Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808354
This item is not returnable.
View return policy
Description
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predomitly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.Specifications
| KLK3 | |
| Monoclonal | |
| Unconjugated | |
| KLK3 | |
| APS, KLK2A1, PSA, hK3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 354 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1A1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07288 | |
| KLK3 | |
| Human recombit protein fragment corresponding to amino acids 25-261 of human KLK3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction