missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK3 Monoclonal Antibody (OTI2C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808357
This item is not returnable.
View return policy
Description
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predomitly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.Specifications
| KLK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07288 | |
| KLK3 | |
| Human recombit protein fragment corresponding to amino acids 25-261 of human KLK3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C9 | |
| Unconjugated | |
| KLK3 | |
| APS, KLK2A1, PSA, hK3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 354 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion