Learn More
Invitrogen™ KLK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109888
Description
Immunogen sequence: QIINGEDCSP HSQPWQAALV MENELFCSGV LVHPQWVLSA AHCFQNSYTI GLGLHSLEAD QEPGSQMVEA SLSVRHPEYN RPLLANDLML IKLDESVSES DTIRSISIAS QCPTAGNSCL VSGWGLLANG RMPTVLQCVN VSVVSEEVCS KLYDPLYHPS MFCAGGGQDQ KDSCNGDSGG PLICNGYLQG LVSFGKAPCG QVGVPGVYTN LCKFTEWIEK TVQAS.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
Specifications
| KLK4 | |
| Polyclonal | |
| Unconjugated | |
| KLK4 | |
| AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56640, 9622 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9Y5K2, Q9Z0M1 | |
| KLK4 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.