Learn More
KLK8 Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801585
Encompass_Preferred
 View more versions of this product
Product Code. 17529942
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
KLK8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60259
KLK8
Human recombit protein fragment corresponding to amino acids 33-260 of human KLK8 produced in SF9 cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3E6
Unconjugated
KLK8
HNP, NP, NRPN, PRSS19, TADG14
Mouse
Affinity Chromatography
RUO
11202
-20° C, Avoid Freeze/Thaw Cycles