Learn More
KLK8 Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801585S
Encompass_Preferred
 View more versions of this product
Product Code. 17549942
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
KLK8 Monoclonal antibody specifically detects KLK8 in Human samples. It is validated for Western Blot
Specifications
Specifications
KLK8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P46013
Mouse
Affinity chromatography
RUO
4288
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3E6
Unconjugated
Mki67
brain serine protease 1; BSP1; EGK_10950; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; neuropsin; NP; Nrpn; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322
Human recombinant protein fragment corresponding to amino acids 1160-1493 of human MKI67 produced in E.coli.
30 μL