missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK8 Monoclonal Antibody (OTI4A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506164S
This item is not returnable.
View return policy
Description
KLK8 Monoclonal antibody specifically detects KLK8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| KLK8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60259 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11202 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A5 | |
| Unconjugated | |
| KLK8 | |
| brain serine protease 1; BSP1; EGK_10950; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; neuropsin; NP; Nrpn; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322 | |
| Full length human recombinant protein of human KLK8 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction