missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK8 Monoclonal Antibody (OTI8F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506134
This item is not returnable.
View return policy
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.Specifications
| KLK8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60259 | |
| KLK8 | |
| Full length human recombit protein of human KLK8 produced in HEk293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI8F2 | |
| Unconjugated | |
| KLK8 | |
| HNP, NP, NRPN, PRSS19, TADG14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11202 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction