missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT12 Monoclonal Antibody (OTI3E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805548S
This item is not returnable.
View return policy
Description
NUDT12 Monoclonal antibody specifically detects NUDT12 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NUDT12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQG2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83594 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E7 | |
| Unconjugated | |
| Nudt12 | |
| 0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12 | |
| Human recombinant protein fragment corresponding to amino acids 1-251 of human NUDT12 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction