missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klotho Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76511AF594
This item is not returnable.
View return policy
Description
Klotho Polyclonal antibody specifically detects Klotho in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Klotho | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| EC 3.2.1, EC 3.2.1.31, klotho | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the center of human Klotho. The immunogen is located within amino acids 410-460 of Klotho. Amino Acid Sequence: SGTTKRRDDAKYMYYLK | |
| 0.1 mL | |
| Signal Transduction | |
| 9365 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction