missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klotho beta Antibody (1025C), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB58891X
This item is not returnable.
View return policy
Description
Klotho beta Monoclonal antibody specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| Klotho beta | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 152831 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 1025C | |
| Flow Cytometry, Immunocytochemistry | |
| beta-klotho, betaKlotho, BKL, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Mouse myeloma cell line, NS0-derived recombinant human Klotho beta with a C-terminal 10 His tag., Phe53-Leu997, Accession # Q86Z14 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction