missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klotho beta Antibody (RG7992), Alexa Fluor™ 532, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28016AF532
This item is not returnable.
View return policy
Description
Klotho beta Monoclonal antibody specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Klotho beta | |
| Monoclonal | |
| Alexa Fluor 532 | |
| beta-klotho, betaKlotho, BKL, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Klotho Beta | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| RG7992 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 152831 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction