missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRC1/2/3 Antibody (20d5), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-28100G
This item is not returnable.
View return policy
Description
KLRC1/2/3 Monoclonal specifically detects KLRC1/2/3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| KLRC1/2/3 | |
| Monoclonal | |
| DyLight 488 | |
| CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse NKG2 (Mr approx. 43 kDa) | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 20d5 | |
| Flow Cytometry | |
| KLRC1 | |
| CHO cells transfected with B6 allele of NKG2A gene | |
| 0.25 mL | |
| Signal Transduction | |
| 3821 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction