missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Antibody (1151A), PE/Atto594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB6944PEATT594
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| KLRG1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1151A | |
| Flow Cytometry | |
| 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
| Chinese hamster ovary cell line CHO-derived mouse KLRG1, Glu57-Tyr188, Accession # O88713 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 10219 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction