missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Golden Hamster, APC, Clone: 2F1, enQuireBio™
Golden Hamster Monoclonal Antibody
Brand: EnquireBio QAB74-APC-25ug
This item is not returnable.
View return policy
Specifications
| KLRG1 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Klrg1 | |
| Protein A/G Purified | |
| RUO | |
| 50928 | |
| IgG |
| Flow Cytometry | |
| 2F1 | |
| APC | |
| NP_058666 | |
| Golden Syrian Hamster | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction