missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10698-1-AP-150UL
This item is not returnable.
View return policy
Description
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.Specifications
| KMO | |
| Polyclonal | |
| Unconjugated | |
| KMO | |
| KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 59113, 8564, 98256 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15229, O88867, Q91WN4 | |
| Kmo | |
| KMO Fusion Protein Ag1109 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction