missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2A/MLL Antibody (BL-175-7E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-76397
This item is not returnable.
View return policy
Description
KMT2A/MLL Monoclonal specifically detects KMT2A/MLL in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| KMT2A/MLL | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 with 0.09% Sodium Azide | |
| KMT2A | |
| Immunogen sequence between amino acids 720 and 780 | |
| 100 μL | |
| 4297.0 | |
| Store at 2-8°C/ 1 year from date of receipt | |
| IgG |
| Western Blot, Immunoprecipitation | |
| BL-175-7E8 | |
| Western Blot 1:1000, Immunoprecipitation 10 μL/0.5 mg lysate | |
| ALL1, ALL-1MLL/GAS7, CXXC7TET1-MLL, CXXC-type zinc finger protein 7, EC 2.1.1.43, HRXFLJ11783, HTRX, HTRX1MLL-AF4 der(11) fusion protein, KMT2ACDK6/MLL fusion protein, Lysine N-methyltransferase 2A, MLL/GAS7 fusion protein, MLL/GMPS fusion protein, MLL1, MLL1A, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog), myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila), Trithorax-like protein, TRX1histone-lysine N-methyltransferase MLL, Zinc finger protein HRX | |
| Rabbit | |
| Antigen Affinity-purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction