missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KMT2C Monoclonal Antibody (1686CT202.60.69)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524750

Product Code. 15954394

  • € 536.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KMT2C Monoclonal Antibody for Western Blot

Histone methyltransferase. Methylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Central component of the MLL2/3 complex, a coactivator complex of nuclear receptors, involved in transcriptional coactivation. KMT2C/MLL3 may be a catalytic subunit of this complex. May be involved in leukemogenesis and developmental disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KMT2C
Monoclonal
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q8NEZ4
KMT2C
human KMT2C recombinant protein.
200 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
1686CT202.60.69
Unconjugated
KMT2C
2.1.1.43; ALR-like protein; HALR; Histone-lysine N-methyltransferase 2C; histone-lysine N-methyltransferase MLL3; histone-lysine N-methyltransferase, H3 lysine-4 specific; homologous to ALR protein; KIAA1506; KMT2C; lysine (K)-specific methyltransferase 2C; lysine methyltransferase 2C; Lysine N-methyltransferase 2C; MLL3; Myeloid/lymphoid or mixed-lineage leukemia protein 3
Mouse
Protein G, Size-exclusion - Dialysis
RUO
58508
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.