missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KMT2C Monoclonal Antibody (7D5D7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548775
This item is not returnable.
View return policy
Description
KMT2C Monoclonal Antibody for Western Blot, Flow, ELISA
Histone methyltransferase. Methylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Central component of the MLL2/3 complex, a coactivator complex of nuclear receptors, involved in transcriptional coactivation. KMT2C/MLL3 may be a catalytic subunit of this complex. May be involved in leukemogenesis and developmental disorder.
Specifications
| KMT2C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8NEZ4 | |
| KMT2C | |
| Purified recombinant fragment of human KMT2C (AA: 1-205) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Western Blot | |
| 7D5D7 | |
| Unconjugated | |
| KMT2C | |
| 2.1.1.43; ALR-like protein; HALR; Histone-lysine N-methyltransferase 2C; histone-lysine N-methyltransferase MLL3; histone-lysine N-methyltransferase, H3 lysine-4 specific; homologous to ALR protein; KIAA1506; KMT2C; lysine (K)-specific methyltransferase 2C; lysine methyltransferase 2C; Lysine N-methyltransferase 2C; MLL3; Myeloid/lymphoid or mixed-lineage leukemia protein 3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 58508 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction