missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-32508
This item is not returnable.
View return policy
Description
KMT2D Polyclonal antibody specifically detects KMT2D in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KMT2D | |
| Polyclonal | |
| Western Blot 1:500-2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| Recombinant protein within C terminal human KMT2D. (Uniprot: O14686) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8085 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction