missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KMT2D Monoclonal Antibody (CL5346)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA535029

Product Code. 17175693

  • € 636.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AQLADTLFSK GLGPWDPPDN LAETQKPEQS SLVPGHLDQV NGQVVPEASQ LSIKQEPREE PCALGAQSVK REANGEPIGA PGTSNHLLLA GPRSEAGHLL LQKLLRAKNV QLSTGRGSEG LRAEINGHI Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 90%.

KMT2D is a histone methyltransferase that methylates Lys-4 of histone H3 (H3K4me). H3K4me represents a specific tag for epigenetic transcriptional activation. KMT2D acts as a coactivator for estrogen receptors by being recruited by ESR1 and activating transcription. KMT2D is involved in chromatin organization, chormatin silencig, oocyte growth, oogenesis, and a positive regulation of cell proliferation. Mutations in the KMT2D gene cause Kabuki syndrome (KABUK1) which results in congential mental retardation syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KMT2D
Monoclonal
1.0 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14686
KMT2D
Recombinant protein fragment of Human KMT2D (Product #RP-102441)
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry
CL5346
Unconjugated
KMT2D
AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21
Mouse
Protein A
RUO
8085
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.