missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KNAT3 Polyclonal Antibody

Description
KNAT3 Polyclonal Antibody for Western Blot, ELISA
KNAT3 (KNOTTED1-like homeobox protein 3) is a gene that encodes a transcription factor protein. Transcription factors are proteins that regulate the expression of other genes by binding to specific DNA sequences and either promoting or inhibiting their transcription. KNAT3 is a member of the KNOX (KNOTTED1-like homeobox) family of transcription factors, which are involved in various developmental processes, including meristem formation and organogenesis. The specific function of KNAT3 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease.
Specifications
Specifications
| Antigen | KNAT3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | KNAT3 |
| Gene Accession No. | P48000 |
| Gene Alias | Homeobox protein knotted-1-like 3KNAT3 At5g25220 F21J6.18 |
| Gene Symbols | KNAT3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?