Learn More
Epredia™ Knife carrier for standard knife carrier ER for long stroke microtomes Epredia™ Microm
Brand: Epredia™ 705820
Additional Details : Weight : 1.00000kg
Specifications
Standard Solid Knife Carrier N | |
Epredia Microm Long Stroke Microtomes (HM 355S, HM 340E, HM 325) | |
Integrated blade guard and clearance angle adjustment; Removable cutting angle set lever |