missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KNOP1 Polyclonal Antibody

Product Code. 15797271 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15797271 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15797271 Supplier Invitrogen™ Supplier No. PA559418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TAGFENEDQK LKFLRLMGGF KNLSPSFSRP ASTIARPNMA LGKKAADSLQ QNLQRDYDRA MSWKYSRGAG LGFSTAPNKI FYIDRNASKS VKL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

KNOP1 is a nucleolar protein that interacts with zinc finger 106 protein. The encoded protein has several of the same characteristics as nucleostemin and may be involved in testis development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNOP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KNOP1
Gene Accession No. Q1ED39
Gene Alias 101F10.1; C16orf88; FAM191A; family with sequence similarity 191, member A; KNOP1; lysine rich nucleolar protein 1; lysine-rich nucleolar protein 1; protein C16orf88; Protein FAM191A; Testis-specific gene 118 protein; TSG118
Gene Symbols KNOP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KNOP1. Recombinant protein control fragment (Product #RP-98633).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.