Description
- Protects the environment from droplets emitted by the wearer
- Effective against bacteria, particles and viruses (BFE, PFE, and VFE)
- Feature exclusive Origami folding and Smile welding for greater comfort and an ideal fit on the wearer's face
- Softex technology limits irritation to the face
- Does not wear out with prolonged use, even in contact with beard
- Hypoallergenic
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| Color | Blue |
| Fastening Type | Earloops |
| Material | Polypropylene |
| Size | One Size |
| Sterility | Non-sterile |
| Style | Pouch |
| Disposable | Single-use |
| Latex Free | Yes |