Learn More
Medicom™ KOLMI Type II Op Air™ Medical Mask
Product Code. 17294343 Shop All Medicom Products
Change view
Click to view available options
:
KOLMI Type II Op Air Medical Mask; Product Type: Medical/Procedural Face Mask
Product Code.
17294343 KOLMI Type II Op Air Medical Mask; Product Type: Medical/Procedural Face Mask
1 options
Product must be used on intact skin only. The reuse or extended use may lead to infection or cross-contamination.
Product Code. 17294343
Brand: Medicom™ M1431130
Only null left
Add to basket
Edge
Add to basket
Product must be used on intact skin only. The reuse or extended use may lead to infection or cross-contamination.
KOLMI Type II Op Air™ Medical Mask with Softex technology provides a soft, anti-irritant inner layer for extra comfort and breathability.
  • Protects the environment from droplets emitted by the wearer
  • Effective against bacteria, particles and viruses (BFE, PFE, and VFE)
  • Feature exclusive Origami folding and Smile welding for greater comfort and an ideal fit on the wearer's face
  • Softex technology limits irritation to the face
  • Does not wear out with prolonged use, even in contact with beard
  • Hypoallergenic
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pouch
Disposable Single-use
Latex Free Yes