Learn More
DWK Life Sciences Kimble™ Kontes™ Brand 3mm NMR Tubes
Product Code. 10059771
Change view
Click to view available options
Length (English):
7 in.
8 in.
Field Strength:
100MHz
300MHz
400MHz
500MHz
600 to 800MHz
Camber (English):
±0.00025 in.
±0.0005 in.
±0.0006 in.
±0.001 in.
±0.0011 in.
±0.002 in.
Concentricity (English):
±0.0005 in.
±0.001 in.
±0.0015 in.
±0.002 in.
±0.0025 in.
Unit Size:
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Field Strength Camber (English) Concentricity (English) unitSize
10059771 7 in. 400MHz ±0.001 in. ±0.0015 in. Pack of 5
10395022 8 in. 500MHz ±0.0006 in. ±0.001 in. Pack of 5
10233252 7 in. 300MHz ±0.0005 in. ±0.002 in. Pack of 5
10542483 7 in. 100MHz ±0.002 in. ±0.0025 in. Pack of 5
10049771 7 in. 500MHz ±0.0005 in. ±0.001 in. Pack of 5
11883133 7 in. 600 to 800MHz ±0.00025 in. ±0.0005 in. Pack of 5
10405162 8 in. 300MHz ±0.0011 in. ±0.002 in. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10059771 Supplier DWK Life Sciences Supplier No. 8978300000
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.
Precision-manufactured tubes designed for use in high-resolution NMR spectroscopy that characterizes organic molecules through measurement of the resonant frequencies of atomic nuclei. Research applications include analysis of polymer structures and organic solutions.
  • Made from Type I borosilicate glass
  • Wall thickness: 0.01118 in. (0.28cm)
  • Wall variation and camber depend, in part, on tube length
  • All sample tubes come with plastic compression-fit caps and sandblasted marking spots
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 0.11 in.
Length (Metric) 177.8 mm
Material Borosilicate Glass
Closure Material Polyethylene
Closure Type Compression-fit
Diameter (Metric) Outer 3 mm
Concentricity (English) ±0.0015 in.
Diameter (English) Inner 0.094 in.
Camber (English) ±0.001 in.
Diameter (Metric) Inner 2.4 mm
Field Strength 400MHz
For Use With (Application) Research applications include analysis of polymer structures and organic solutions
Includes Cap
Length (English) 7 in.
Shape Cylindrical
Thickness (English) Wall 0.011 in.
Thickness (Metric) Wall 0.28 mm