Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Product Code. 10248244
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
200 mm
300 mm
1200 mm
Column Capacity:
25 mL
49 mL
74 mL
98 mL
147 mL
638 mL
Quantity:
2/Cs.
5/Cs.
Each
Unit Size:
Each
Pack of 2
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Column Capacity Quantity Unit Size
10248244 1200 mm 638 mL 2/Cs. Pack of 2
11772364 100 mm 49 mL 5/Cs. Pack of 5
11782364 200 mm 98 mL 5/Cs. Pack of 5
12943351 100 mm 49 mL Each Each
12973351 150 mm 74 mL 5/Cs. Pack of 5
12913361 300 mm 147 mL 5/Cs. Pack of 5
12933351 50 mm 25 mL 5/Cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10248244 Supplier DWK Life Sciences Supplier No. 4204002512
  • ONLINE EXCLUSIVE PRICE
    € 336.00
    € 256.92 / Pack of 2
    Save € 79.08
    23% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • With flow adapters
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Specifications
Max. Pressure 14.5 psi
Pore Size 20 μm
Column Capacity 638 mL
Length (Metric) 1200 mm
Material Borosilicate Glass Type I Class A
Support Material HDPE
Connector Types Male luer lock
Cross Sectional Area 4.91cm3
Sterilization Method 2N NaOH or 100% Ethanol
Temperature (Metric) Range 0 to 50°C
Quantity 2/Cs.
For Use With (Application) Ion exchange, affinity, gel filtration and hydrophobic interaction media
Endcapped Yes
Type Economy Chromatography Column
Diameter (Metric) Inner 25 mm