Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Borosilicate Glass Thrift N-51A 5mm NMR Tubes
Product Code. 12623195
Click to view available options
Length (English):
8 in.
Camber (English):
0.00035 in.
Concentricity (English):
0.0005 in.
Field Strength:
400MHz
Unit Size:
Pack of 5
Product Code. 12623195
Brand: DWK Life Sciences 8971400008
Only null left
Add to basket
Add to basket
Includes
Case of 1
Used to contain samples in nuclear magnetic resonance spectroscopy.
Used to contain samples in nuclear magnetic resonance spectroscopy
  • Made of KIMAX N-51A borosilicate glass
  • Have attached cap
  • Wall thickness is 0.014 in. (0.38mm)
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 0.19 in.
Length (Metric) 203.2 mm
Material Borosilicate Glass
Closure Material Polyethylene
Diameter (Metric) Outer 5 mm
Concentricity (English) 0.0005 in.
Camber (English) 0.00035 in.
Field Strength 400MHz
For Use With (Application) Used to contain samples in nuclear magnetic resonance spectroscopy
Includes Cap
Length (English) 8 in.
Thickness (English) Wall 0.014 in.
Thickness (Metric) Wall 0.38 mm
Quantity 1/Cs.