Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ NMR Borosilicate Glass 5mm Tube
Product Code. 10641713
Click to view available options
Length (English):
7 in.
8 in.
Camber (English):
0.00025 in.
0.00035 in.
Unit Size:
Pack of 5
Product Code. 10641713
Brand: DWK Life Sciences 8972410008
Only null left
Add to basket
Add to basket
Ideal for sealing directly to vacuum manifolds, joints or valves
  • Made of KIMAX KG-33 borosilicate glass
  • Have attached polyethylene cap
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 0.19 in.
Length (Metric) 203.2 mm
Material Borosilicate Glass
Diameter (Metric) Outer 5 mm
Concentricity (English) 0.0005 in.
Diameter (English) Inner 0.16 in.
Camber (English) 0.00035 in.
Diameter (Metric) Inner 4.2 mm
Field Strength 600 to 700 MHz
For Use With (Application) Ideal for sealing directly to vacuum manifolds, joints or valves
Includes Cap
Length (English) 8 in.
Thickness (English) Wall 0.014 in.
Thickness (Metric) Wall 0.38 mm