missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes

Product Code. 12603205
Change view
Click to view available options
Color:
Black
Green
Red
White
Yellow
For Use With (Equipment):
5 mm NMR Sample Tube
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) unitSize
12603205 Black 5 mm NMR Sample Tube Pack of 100
12633205 Yellow 5 mm NMR Sample Tube Pack of 100
12613205 White 5 mm NMR Sample Tube Pack of 100
11873143 Red 5 mm NMR Sample Tube Pack of 100
12623205 Green 5 mm NMR Sample Tube Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12603205 Supplier DWK Life Sciences Supplier No. 8970950021
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures

TRUSTED_SUSTAINABILITY

Specifications

Product Type Pressure Cap
Material Polyethylene
Color Black
For Use With (Application) Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
For Use With (Equipment) 5 mm NMR Sample Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.