Learn More
DWK Life Sciences Kimble™ Kontes™ Neoprene Gasket for NMR Tube Washer
Product Code. 15122706
Change view
Click to view available options
For Use With (Application):
For NMR utility washer
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
15122706 For NMR utility washer Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15122706 Supplier DWK Life Sciences Supplier No. 4599510000
Replacement gasket for utility washers.
  • Utility washers are used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items
  • Unit is supplied with one gasket, a tube washer, an Erlenmeyer flask and a 3/8 in. hose connection
  • Replacement gaskets, Case of 12
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For NMR utility washer
Material Neoprene
Type Gasket