Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Micro Syringes
DWK Life Sciences Kimble&trade Kontes&trade Micro Syringes feature a Luer slip tip and are suitable for work in chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
Brand: DWK Life Sciences 748019-0001
13.35 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Polypropylene barrel
- Polyethylene plunger
- Latex free
- More chemically resistant than rubber-tipped syringes
- For research use only; not for use in diagnostic procedures
- Sterile, individually wrapped needles contain no rubber, silicone oil, styrene, DNA, latex, or DEHP
Specifications
Polyethylene | |
Microsyringe | |
0.01 cc | |
1 mL |