Learn More
DWK Life Sciences Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Product Code. 16444320
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16444320 50 mL Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16444320 Supplier DWK Life Sciences Supplier No. KC4506625150
Only null left
Add to basket
Add to basket
Uniform wall thickness for maximum heat transfer and heat resistance
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
  • KG-33 borosilicate glass
  • Designed from ASTM™ E982, Type VI, Class A requirements
  • Thread finish complies with Glass Packaging Institute (GPI) requirements
  • Cap is not attached
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E 438 Type I, Class A
Closure Material Phenolic
Capacity (Metric) 50 mL
Length (Metric) 150 mm
Material Borosilicate Glass
Disposable Reusable
For Use With Biohazard test protocols
Includes Unattached cap
Thread Size 24-410
Diameter (Metric) 25 mm
Closure Type Screw Cap