Få mere at vide
Få mere at vide
DWK Life Sciences Kimble™ Kontes™ Squibb Pear-Shaped Separatory Funnels with standard taper Stopper and Autoclavable PTFE Stockcock Plug
Klik for at se tilgængelige muligheder
Capacity (Metric):
125 mL
1000 mL
Stopcock:
2; PTFE
4; PTFE
Quantity:
1 Case
4 Case
Pakningsstørrelse:
Each
Pack of 4
Beskrivelse
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- Lower stems have inside diameter large enough so that a column of liquid will break with the stopcock closed, thereby giving a more complete separation and eliminating the necessity of emptying the funnel
- Designed from ASTM™ Specifications E1096, Type 4 requirements
Tekniske data
Tekniske data
| Capacity (Metric) | 125 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E1096, Type IV |
| Length (Metric) Stem | 60mm |
| Quantity | 4 Case |
| Stopcock | 2; PTFE |
| Stopper Material | Glass |
| Product Type | Squibb Separatory Funnel |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion