Learn More
KOR Antibody (387301), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3895H
Product Code. 18645053
  • € 652.00 / 0.10mL
Description
KOR Monoclonal antibody specifically detects KOR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KOR
Monoclonal
HRP
PBS
Mouse
0.1 mL
GPCR, Phospho Specific, Signal Transduction
4986
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, CyTOF
387301
Flow Cytometry, Immunocytochemistry, CyTOF-ready
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK
NS0 mouse myeloma cell line transfected with human KOR, Met1-Val380, Accession # P41145