missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KOR Antibody (387301), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3895J
This item is not returnable.
View return policy
Description
KOR Monoclonal antibody specifically detects KOR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| KOR | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 387301 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
| NS0 mouse myeloma cell line transfected with human KOR, Met1-Val380, Accession # P41145 | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 4986 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction