missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kos1/Tnk1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-237DL594
This item is not returnable.
View return policy
Description
Kos1/Tnk1 Polyclonal antibody specifically detects Kos1/Tnk1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Kos1/Tnk1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| C-terminal residues 421-434 [QNQVQSLPAGSAEP] of the 47 kDa mouse Kos1/ Tnk1 protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 594 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction