Learn More
KPNA3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28050-1-AP-150UL
 View more versions of this product
Product Code. 18018965
  • € 410.00 / 150µL
Description
The KPNA3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KPNA3. This antibody recognizes human, mouse antigen. The KPNA3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
KPNA3
650 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
hSRP1, Importin alpha Q2, Importin subunit alpha 3, IPOA4, Karyopherin subunit alpha 3, KPNA3, Qip2, SRP1, SRP1 gamma, SRP1gamma, SRP4
150 μL
3839
Store at -20°C. Stable for one year after shipment.