Learn More
Invitrogen™ KPNA6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596654
Description
Immunogen sequence: SYLSDGPNEK IQAVIDSGVC RRLVELLMHN DYKVASPALR AVGNIVTGDD IQTQVILNCS ALPCLLHLLS SPKESIRKEA CWTISNITAG NRAQIQAVID ANIFPVLIEI LQKAEFRTRK EAAWAITNAT SGGTPEQIRY LVSLGCIKPL CDLLTVMDSK IVQVALNGLE NILRLGEQEG KRSGSGVNPY CGLIEEAYGL DKIEFLQSHE NQEIYQKAFD LIEHYFGVED DDSSLAPQVD ETQQQFIFQQ PEAPMEGFQL; Positive Samples: BT-474, SW620, U-251MG, Mouse testis.
Specifications
| KPNA6 | |
| Polyclonal | |
| Unconjugated | |
| KPNA6 | |
| FLJ11249; importin alpha 7 subunit; importin alpha-S2; Importin subunit alpha-7; importin-alpha-S2; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 6; karyopherin alpha 6 (importin alpha 7); karyopherin subunit alpha 6; karyopherin subunit alpha-6; Kpna5; Kpna6; KPNA7; MGC17918; NPI-2; RP4-622L5.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16650, 23633 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35345, O60684 | |
| KPNA6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6 (NP_0364481). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.