missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | KPNA6 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315796
|
Novus Biologicals
NBP3-04919-20UL |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394535
|
Novus Biologicals
NBP3-04919-100UL |
100 μL |
€ 470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KPNA6 Polyclonal specifically detects KPNA6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| KPNA6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| importin alpha 7 subunit, importin subunit alpha-7, importin-alpha-S2, IPOA7FLJ11249, karyopherin alpha 6 (importin alpha 7), Karyopherin subunit alpha-6, KPNA7, MGC17918 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6 (NP_036448.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 23633 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title