missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA7 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81730JF549
This item is not returnable.
View return policy
Description
KPNA7 Polyclonal antibody specifically detects KPNA7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| KPNA7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 402569 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| karyopherin alpha 7 (importin alpha 8) | |
| KPNA7 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of human KPNA7. The immunogen is located within amino acids 40 - 90 of KPNA7. Amino Acid Squence: PDTPSEKTAKGV | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction