missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNB1 Monoclonal Antibody (OTI10D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811700S
This item is not returnable.
View return policy
Description
KPNB1 Monoclonal antibody specifically detects KPNB1 in Human samples. It is validated for Western BlotSpecifications
| KPNB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75052 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9722 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI10D3 | |
| Unconjugated | |
| NOS1AP | |
| AA409963; HGNC:6400; IMB1; Impnb; importin 1; importin 90; importin beta; importin beta-1 subunit; importin subunit beta-1; Importin-90; IPO1; IPOB; KapB1; karyopherin (importin) beta 1; karyopherin beta 1; karyopherin subunit beta 1; karyopherin subunit beta-1; karyopherin,beta 1; Kpnb1; MGC2155; MGC2156; MGC2157; NTF97; Nuclear factor p97; pore targeting complex 97 kDa subunit; PTAC97; SCG | |
| Human recombinant protein fragment corresponding to amino acids 1-196 of human NOS1AP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction