missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KPRP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546028
This item is not returnable.
View return policy
Description
Peptide sequence: PHRLDTEAPY CGPSSYNQGQ ESGAGCGPGD VFPERRGQDG HGDQGNAFAG Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| KPRP | |
| Polyclonal | |
| Unconjugated | |
| KPRP | |
| C1orf45; hKPRP; keratinocyte expressed, proline-rich protein; keratinocyte proline rich protein; keratinocyte proline-rich protein; KPRP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 448834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5T749 | |
| KPRP | |
| Synthetic peptide directed towards the C-terminal of human KPRP (aa 520-569). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction